The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198321.3 | NP_938080.1 | 8 - 9 | 1294 | 70 |
GenBank mRNA | AB078145.1 | - | 8 - 9 | 1162 | 70 |
AJ505950.1 | - | 8 - 9 | 1162 | 70 | |
AK023782.1 | - | 5 - 6 | 628 | 70 | |
AK127135.1 | - | 8 - 9 | 1263 | 70 | |
BC007224.1 | - | 1 - 2 | 94 | 70 | |
BC072450.1 | - | 4 - 5 | 594 | 70 | |
BC098444.1 | - | 3 - 4 | 283 | 70 |