The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145211.2 | NP_001138683.1 | 6 - 7 | 869 | 64 |
NM_001145212.2 | NP_001138684.1 | 3 - 4 | 742 | 64 | |
NM_007256.4 | NP_009187.1 | 6 - 7 | 1174 | 64 | |
XM_017017157.1 | XP_016872646.1 | 6 - 7 | 792 | 64 | |
GenBank mRNA | AB020687.1 | - | 6 - 7 | 1045 | 64 |
AB026256.1 | - | 6 - 7 | 957 | 64 | |
AF205073.1 | - | 6 - 7 | 891 | 64 | |
AK290234.1 | - | 6 - 7 | 1056 | 64 | |
AK294503.1 | - | 6 - 7 | 854 | 64 | |
AK300134.1 | - | 3 - 4 | 614 | 64 | |
AK302492.1 | - | 4 - 5 | 819 | 64 | |
BC041095.1 | - | 6 - 7 | 1043 | 64 |