Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
U2SURP
Entrez Gene ID
23350
Gene Name
U2 snRNP associated SURP domain containing
Gene Aliases
SR140, fSAPa
Chromosome Location
Chr.3: 143001524 - 143060725 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080415.1 NP_001073884.1 7 - 8 741 176
NM_001320219.1 NP_001307148.1 7 - 8 741 176
NM_001320220.1 NP_001307149.1 7 - 8 741 176
NM_001320222.1 NP_001307151.1 7 - 8 741 176
XM_005247248.4 XP_005247305.1 7 - 8 747 176
XM_017006038.1 XP_016861527.1 7 - 8 680 176
GenBank mRNA AB002330.1 - 7 - 8 640 176
AK296440.1 - 7 - 8 661 176
BC006474.1 - 7 - 8 640 176
BC111692.1 - 7 - 8 1112 176

View Full Product Details