Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD6
Entrez Gene ID
9861
Gene Name
proteasome 26S subunit, non-ATPase 6
Gene Aliases
Rpn7, S10, SGA-113M, p42A, p44S10
Chromosome Location
Chr.3: 64010549 - 64024010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014814.2 NP_055629.1 1 - 2 335 71
GenBank mRNA AF215935.1 - 1 - 2 201 71
AF530062.1 - 1 - 2 148 71
AK290205.1 - 1 - 2 189 71
AY568086.1 - 1 - 2 173 71
BC000630.1 - 1 - 2 165 71
BC000904.2 - 1 - 2 186 71
BC012369.1 - 1 - 2 183 71
D14663.1 - 1 - 2 173 71
HY010377.1 - 1 - 2 336 71

View Full Product Details