The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271779.1 | NP_001258708.1 | 3 - 4 | 689 | 65 |
NM_001271780.1 | NP_001258709.1 | 2 - 3 | 404 | 65 | |
NM_001271781.1 | NP_001258710.1 | 2 - 3 | 279 | 65 | |
NM_014814.2 | NP_055629.1 | 2 - 3 | 530 | 65 | |
XM_005265619.1 | XP_005265676.1 | 2 - 3 | 626 | 65 | |
XM_011534288.1 | XP_011532590.1 | 3 - 4 | 585 | 65 | |
XM_017007569.1 | XP_016863058.1 | 3 - 4 | 432 | 65 | |
GenBank mRNA | AF215935.1 | - | 2 - 3 | 396 | 65 |
AF530062.1 | - | 2 - 3 | 343 | 65 | |
AK095059.1 | - | 1 - 2 | 1112 | 65 | |
AK290205.1 | - | 2 - 3 | 384 | 65 | |
AL529900.3 | - | 2 - 3 | 265 | 65 | |
AY359879.1 | - | 2 - 3 | 376 | 65 | |
AY568086.1 | - | 2 - 3 | 368 | 65 | |
BC000630.1 | - | 2 - 3 | 360 | 65 | |
BC000904.2 | - | 2 - 3 | 381 | 65 | |
BC012369.1 | - | 2 - 3 | 378 | 65 | |
BG723464.1 | - | 2 - 3 | 407 | 65 | |
BX647742.1 | - | 3 - 4 | 556 | 65 | |
CD692691.1 | - | 3 - 4 | 589 | 65 | |
D14663.1 | - | 2 - 3 | 368 | 65 |