The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271779.1 | NP_001258708.1 | 4 - 5 | 835 | 126 |
NM_001271780.1 | NP_001258709.1 | 3 - 4 | 550 | 126 | |
NM_001271781.1 | NP_001258710.1 | 3 - 4 | 425 | 126 | |
NM_014814.2 | NP_055629.1 | 3 - 4 | 676 | 126 | |
XM_005265619.1 | XP_005265676.1 | 3 - 4 | 772 | 126 | |
XM_011534288.1 | XP_011532590.1 | 4 - 5 | 731 | 126 | |
XM_017007569.1 | XP_016863058.1 | 4 - 5 | 578 | 126 | |
GenBank mRNA | AF215935.1 | - | 3 - 4 | 542 | 126 |
AF530062.1 | - | 3 - 4 | 489 | 126 | |
AK095059.1 | - | 2 - 3 | 1258 | 126 | |
AK290205.1 | - | 3 - 4 | 530 | 126 | |
AL529900.3 | - | 3 - 4 | 411 | 126 | |
AY568086.1 | - | 3 - 4 | 514 | 126 | |
BC000630.1 | - | 3 - 4 | 506 | 126 | |
BC000904.2 | - | 3 - 4 | 527 | 126 | |
BC012369.1 | - | 3 - 4 | 524 | 126 | |
BX647742.1 | - | 4 - 5 | 702 | 126 | |
D14663.1 | - | 3 - 4 | 514 | 126 |