Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMD6
Entrez Gene ID
9861
Gene Name
proteasome 26S subunit, non-ATPase 6
Gene Aliases
Rpn7, S10, SGA-113M, p42A, p44S10
Chromosome Location
Chr.3: 64010549 - 64024010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271779.1 NP_001258708.1 5 - 6 1064 73
NM_001271780.1 NP_001258709.1 4 - 5 779 73
NM_001271781.1 NP_001258710.1 4 - 5 654 73
NM_014814.2 NP_055629.1 4 - 5 905 73
XM_005265619.1 XP_005265676.1 4 - 5 1001 73
XM_011534288.1 XP_011532590.1 5 - 6 960 73
XM_017007569.1 XP_016863058.1 5 - 6 807 73
GenBank mRNA AF215935.1 - 4 - 5 771 73
AF530062.1 - 4 - 5 718 73
AK095059.1 - 3 - 4 1487 73
AK290205.1 - 4 - 5 759 73
AL529900.3 - 4 - 5 640 73
AY359879.1 - 4 - 5 751 73
AY568086.1 - 4 - 5 743 73
BC000630.1 - 4 - 5 735 73
BC000904.2 - 4 - 5 756 73
BC012369.1 - 4 - 5 753 73
BX647742.1 - 5 - 6 931 73
D14663.1 - 4 - 5 743 73

View Full Product Details