Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FGD4
- Entrez Gene ID
-
121512
- Gene Name
- FYVE, RhoGEF and PH domain containing 4
- Gene Aliases
-
CMT4H, FRABP, ZFYVE6
- Chromosome Location
-
Chr.12: 32399523 - 32646050 on Build GRCh38
View Full Product Details