Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FGD4
Entrez Gene ID
121512
Gene Name
FYVE, RhoGEF and PH domain containing 4
Gene Aliases
CMT4H, FRABP, ZFYVE6
Chromosome Location
Chr.12: 32399523 - 32646050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304480.1 NP_001291409.1 9 - 10 1643 147
NM_001304481.1 NP_001291410.1 9 - 10 1609 147
NM_001304483.1 NP_001291412.1 10 - 11 1833 147
NM_001304484.1 NP_001291413.1 10 - 11 1852 147
NM_139241.3 NP_640334.2 9 - 10 1733 147
XM_005253304.4 XP_005253361.1 9 - 10 1869 147
XM_005253307.3 XP_005253364.1 8 - 9 1845 147
XM_005253308.4 XP_005253365.1 8 - 9 1897 147
XM_005253309.1 XP_005253366.1 8 - 9 1357 147
XM_005253310.3 XP_005253367.1 5 - 6 527 147
XM_011520554.1 XP_011518856.1 9 - 10 1517 147
XM_011520555.1 XP_011518857.1 9 - 10 1559 147
XM_011520556.1 XP_011518858.1 10 - 11 1788 147
XM_011520557.1 XP_011518859.1 7 - 8 951 147
XM_011520558.2 XP_011518860.1 6 - 7 671 147
XM_011520559.2 XP_011518861.1 5 - 6 513 147
XM_017018803.1 XP_016874292.1 9 - 10 1869 147
XM_017018804.1 XP_016874293.1 8 - 9 1697 147
XM_017018805.1 XP_016874294.1 7 - 8 847 147
GenBank mRNA AK057294.1 - 9 - 10 1616 147
AK297025.1 - 9 - 10 1609 147
AK303334.1 - 9 - 10 1614 147
AY367054.1 - 7 - 8 781 147

View Full Product Details