Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF9
Entrez Gene ID
64147
Gene Name
kinesin family member 9
Gene Aliases
-
Chromosome Location
Chr.3: 47228026 - 47282847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134878.1 NP_001128350.1 12 - 13 1732 122
NM_022342.4 NP_071737.1 11 - 12 1486 122
NM_182902.3 NP_878905.2 11 - 12 1486 122
XM_005265388.4 XP_005265445.1 11 - 12 1261 122
XM_005265389.4 XP_005265446.1 11 - 12 1261 122
XM_006713291.3 XP_006713354.1 10 - 11 1036 122
XM_011534003.2 XP_011532305.1 12 - 13 1461 122
XM_011534004.2 XP_011532306.1 12 - 13 1685 122
XM_011534005.2 XP_011532307.1 10 - 11 993 122
XM_011534006.2 XP_011532308.1 10 - 11 1379 122
XM_011534007.2 XP_011532309.1 4 - 5 260 122
XM_017007029.1 XP_016862518.1 12 - 13 1457 122
XM_017007030.1 XP_016862519.1 11 - 12 1460 122
XM_017007031.1 XP_016862520.1 10 - 11 993 122
GenBank mRNA AF311212.1 - 11 - 12 1189 122
AK023894.1 - 11 - 12 1189 122
AK292547.1 - 11 - 12 1180 122
AK302070.1 - 10 - 11 1130 122
AK302974.1 - 12 - 13 1360 122
AK314418.1 - 12 - 13 1463 122
BC030657.2 - 11 - 12 1237 122

View Full Product Details