Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF9
Entrez Gene ID
64147
Gene Name
kinesin family member 9
Gene Aliases
-
Chromosome Location
Chr.3: 47228026 - 47282847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134878.1 NP_001128350.1 16 - 17 2119 65
NM_182902.3 NP_878905.2 15 - 16 1873 65
XM_005265388.4 XP_005265445.1 15 - 16 1648 65
XM_006713291.3 XP_006713354.1 14 - 15 1423 65
XM_011534003.2 XP_011532305.1 16 - 17 1848 65
XM_011534004.2 XP_011532306.1 16 - 17 2072 65
XM_011534005.2 XP_011532307.1 14 - 15 1380 65
XM_011534006.2 XP_011532308.1 14 - 15 1766 65
XM_011534007.2 XP_011532309.1 8 - 9 647 65
XM_017007029.1 XP_016862518.1 16 - 17 1844 65
XM_017007030.1 XP_016862519.1 15 - 16 1847 65
GenBank mRNA AK023894.1 - 15 - 16 1576 65
AK292547.1 - 15 - 16 1567 65
AK314418.1 - 16 - 17 1850 65
BC030657.2 - 15 - 16 1624 65

View Full Product Details