The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318798.1 | NP_001305727.1 | 8 - 9 | 840 | 88 |
NM_001318800.1 | NP_001305729.1 | 8 - 9 | 1124 | 88 | |
NM_004186.4 | NP_004177.3 | 9 - 10 | 1101 | 88 | |
XM_005265381.4 | XP_005265438.1 | 9 - 10 | 1231 | 88 | |
XM_005265382.4 | XP_005265439.1 | 8 - 9 | 941 | 88 | |
XM_006713289.3 | XP_006713352.1 | 3 - 4 | 713 | 88 | |
XM_006713290.3 | XP_006713353.1 | 9 - 10 | 1034 | 88 | |
XM_011533998.2 | XP_011532300.1 | 9 - 10 | 1033 | 88 | |
XM_011534000.2 | XP_011532302.1 | 9 - 10 | 1034 | 88 | |
GenBank mRNA | AB209259.1 | - | 8 - 9 | 794 | 88 |
BC042914.1 | - | 9 - 10 | 1097 | 88 | |
U33920.1 | - | 8 - 9 | 977 | 88 | |
U38276.1 | - | 8 - 9 | 1118 | 88 |