Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HPS4
Entrez Gene ID
89781
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
Chromosome Location
Chr.22: 26443613 - 26483863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022081.5 NP_071364.4 6 - 7 1167 117
NM_152841.2 NP_690054.1 4 - 5 784 117
XM_006724353.3 XP_006724416.1 6 - 7 1158 117
XM_006724354.3 XP_006724417.1 6 - 7 1063 117
XM_017029042.1 XP_016884531.1 6 - 7 1063 117
XM_017029043.1 XP_016884532.1 7 - 8 1268 117
XM_017029044.1 XP_016884533.1 6 - 7 1066 117
XM_017029046.1 XP_016884535.1 6 - 7 1158 117
XM_017029048.1 XP_016884537.1 6 - 7 1158 117
XM_017029049.1 XP_016884538.1 6 - 7 1063 117
XM_017029050.1 XP_016884539.1 6 - 7 1158 117
XM_017029051.1 XP_016884540.1 6 - 7 1063 117
XM_017029055.1 XP_016884544.1 5 - 6 1067 117
XM_017029057.1 XP_016884546.1 6 - 7 1158 117
XM_017029059.1 XP_016884548.1 6 - 7 1067 117
XM_017029060.1 XP_016884549.1 6 - 7 1066 117
XM_017029062.1 XP_016884551.1 5 - 6 1067 117
XM_017029064.1 XP_016884553.1 5 - 6 950 117
GenBank mRNA AB051454.1 - 6 - 7 1070 117
AK054717.1 - 4 - 5 591 117
AK057648.1 - 6 - 7 996 117
AK290211.1 - 6 - 7 1087 117
AL365512.1 - 4 - 5 688 117
AL713795.1 - 6 - 7 1116 117
AY043416.1 - 5 - 6 687 117
BC065030.1 - 5 - 6 889 117
CR456392.1 - 5 - 6 588 117

View Full Product Details