Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HPS4
- Entrez Gene ID
-
89781
- Gene Name
- HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
- Gene Aliases
-
BLOC3S2, LE
- Chromosome Location
-
Chr.22: 26443613 - 26483863 on Build GRCh38
View Full Product Details