The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014661.3 | NP_055476.3 | 4 - 5 | 1424 | 70 |
GenBank mRNA | AK127343.1 | - | 4 - 5 | 1618 | 70 |
D50930.1 | - | 4 - 5 | 1113 | 70 |