Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDGFRP2
Entrez Gene ID
84717
Gene Name
hepatoma-derived growth factor-related protein 2
Gene Aliases
HDGF-2, HDGF2, HRP-2
Chromosome Location
Chr.19: 4472196 - 4502220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001520.2 NP_001001520.1 3 - 4 446 96
NM_032631.3 NP_116020.1 3 - 4 446 96
XM_011528375.2 XP_011526677.1 3 - 4 806 96
XM_011528376.2 XP_011526678.1 3 - 4 806 96
XM_011528377.2 XP_011526679.1 3 - 4 806 96
GenBank mRNA AB208892.1 - 1 - 2 145 96
AF294267.1 - 3 - 4 353 96
AL520441.3 - 3 - 4 391 96
AY358600.1 - 3 - 4 355 96
BC000755.2 - 3 - 4 336 96
BI826321.1 - 3 - 4 409 96
BI905780.1 - 3 - 4 383 96
BM147540.1 - 3 - 4 381 96

View Full Product Details