The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321431.1 | NP_001308360.1 | 32 - 33 | 4229 | 109 |
NM_032447.4 | NP_115823.3 | 32 - 33 | 4504 | 109 | |
XM_017027372.1 | XP_016882861.1 | 32 - 33 | 4228 | 109 | |
XM_017027373.1 | XP_016882862.1 | 32 - 33 | 4487 | 109 | |
XM_017027374.1 | XP_016882863.1 | 31 - 32 | 4391 | 109 | |
XM_017027375.1 | XP_016882864.1 | 32 - 33 | 4487 | 109 | |
XM_017027376.1 | XP_016882865.1 | 32 - 33 | 4487 | 109 | |
XM_017027377.1 | XP_016882866.1 | 13 - 14 | 1554 | 109 | |
XM_017027378.1 | XP_016882867.1 | 5 - 6 | 781 | 109 | |
XM_017027379.1 | XP_016882868.1 | 33 - 34 | 4613 | 109 | |
GenBank mRNA | AB053450.2 | - | 31 - 32 | 4110 | 109 |
AY165863.1 | - | 32 - 33 | 4504 | 109 | |
AY165864.1 | - | 32 - 33 | 4229 | 109 | |
AY165865.1 | - | 31 - 32 | 4375 | 109 | |
BC153882.1 | - | 31 - 32 | 4110 | 109 | |
CN369190.1 | - | 4 - 5 | 448 | 109 |