Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
Chromosome Location
Chr.19: 8065402 - 8149846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321431.1 NP_001308360.1 45 - 46 5791 65
NM_032447.4 NP_115823.3 45 - 46 6066 65
XM_017027372.1 XP_016882861.1 45 - 46 5790 65
XM_017027373.1 XP_016882862.1 45 - 46 6049 65
XM_017027374.1 XP_016882863.1 44 - 45 5953 65
XM_017027375.1 XP_016882864.1 44 - 45 5926 65
XM_017027377.1 XP_016882866.1 26 - 27 3116 65
XM_017027378.1 XP_016882867.1 18 - 19 2343 65
XM_017027379.1 XP_016882868.1 46 - 47 6175 65
GenBank mRNA AB053450.2 - 44 - 45 5672 65
AK131282.1 - 3 - 4 374 65
AY165863.1 - 45 - 46 6066 65
AY165864.1 - 45 - 46 5791 65
AY165865.1 - 44 - 45 5937 65
BC153882.1 - 44 - 45 5672 65

View Full Product Details