Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
Chromosome Location
Chr.19: 8065402 - 8149846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321431.1 NP_001308360.1 60 - 61 7720 110
NM_032447.4 NP_115823.3 60 - 61 7995 110
XM_017027372.1 XP_016882861.1 60 - 61 7719 110
XM_017027373.1 XP_016882862.1 60 - 61 7918 110
XM_017027374.1 XP_016882863.1 59 - 60 7882 110
XM_017027375.1 XP_016882864.1 59 - 60 7855 110
XM_017027376.1 XP_016882865.1 59 - 60 7849 110
XM_017027377.1 XP_016882866.1 41 - 42 5045 110
XM_017027378.1 XP_016882867.1 33 - 34 4272 110
XM_017027379.1 XP_016882868.1 61 - 62 8104 110
GenBank mRNA AB053450.2 - 59 - 60 7601 110
AK095563.1 - 4 - 5 487 110
AK096539.1 - 15 - 16 1922 110
AY165863.1 - 60 - 61 7995 110
AY165864.1 - 60 - 61 7720 110
AY165865.1 - 59 - 60 7866 110
BC153882.1 - 59 - 60 7601 110

View Full Product Details