Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GHSR
Entrez Gene ID
2693
Gene Name
growth hormone secretagogue receptor
Gene Aliases
GHDP
Chromosome Location
Chr.3: 172443291 - 172448456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004122.2 NP_004113.1 1 - 1 858 65
GenBank mRNA BC069068.1 - 1 - 1 815 65
BC069374.1 - 1 - 1 815 65
U60181.1 - 1 - 1 815 65

View Full Product Details