The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282409.1 | NP_001269338.1 | 5 - 6 | 1307 | 154 |
NM_001282410.1 | NP_001269339.1 | 6 - 7 | 1436 | 154 | |
NM_001282411.1 | NP_001269340.1 | 5 - 6 | 1307 | 154 | |
NM_001282412.1 | NP_001269341.1 | 6 - 7 | 682 | 154 | |
NM_001282413.1 | NP_001269342.1 | 5 - 6 | 553 | 154 | |
NM_001282414.1 | NP_001269343.1 | 5 - 6 | 553 | 154 | |
NM_002712.2 | NP_002703.1 | 6 - 7 | 1436 | 154 | |
XM_011511388.1 | XP_011509690.1 | 5 - 6 | 651 | 154 | |
GenBank mRNA | AK294043.1 | - | 5 - 6 | 508 | 154 |
AK308015.1 | - | 6 - 7 | 648 | 155 | |
BC000910.2 | - | 6 - 7 | 609 | 154 | |
BC012397.1 | - | 6 - 7 | 602 | 154 | |
BC013001.2 | - | 6 - 7 | 614 | 154 | |
BG684153.1 | - | 5 - 6 | 479 | 154 | |
BI256145.1 | - | 5 - 6 | 467 | 154 | |
BQ212238.1 | - | 5 - 6 | 534 | 154 | |
BT007296.1 | - | 6 - 7 | 598 | 154 | |
BT020134.1 | - | 6 - 7 | 598 | 154 | |
Z50749.1 | - | 6 - 7 | 613 | 154 |