The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282409.1 | NP_001269338.1 | 7 - 8 | 1526 | 124 |
NM_001282412.1 | NP_001269341.1 | 8 - 9 | 901 | 124 | |
NM_001282413.1 | NP_001269342.1 | 7 - 8 | 772 | 124 | |
NM_002712.2 | NP_002703.1 | 8 - 9 | 1655 | 124 | |
XM_011511388.1 | XP_011509690.1 | 7 - 8 | 870 | 124 | |
GenBank mRNA | AK294043.1 | - | 7 - 8 | 727 | 124 |
BC000910.2 | - | 8 - 9 | 828 | 124 | |
BT020134.1 | - | 8 - 9 | 817 | 124 | |
Z50749.1 | - | 8 - 9 | 832 | 124 |