Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
Chromosome Location
Chr.1: 228100727 - 228103462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024319.3 NP_077295.1 3 - 4 527 77
NR_130975.1 3 - 4 527 77
GenBank mRNA AK123377.1 - 3 - 4 507 77
AK126087.1 - 3 - 4 2476 77
AY137773.1 - 3 - 4 388 77
BC002843.2 - 3 - 4 386 77
BC062585.1 - 3 - 4 383 77

View Full Product Details