Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VIL1
Entrez Gene ID
7429
Gene Name
villin 1
Gene Aliases
D2S1471, VIL
Chromosome Location
Chr.2: 218419115 - 218449525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007127.2 NP_009058.2 11 - 12 1264 72
GenBank mRNA AK223398.1 - 11 - 12 1258 72
AK300965.1 - 6 - 7 495 72
AK313709.1 - 11 - 12 1256 72
X12901.1 - 10 - 11 1223 72

View Full Product Details