Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VIL1
Entrez Gene ID
7429
Gene Name
villin 1
Gene Aliases
D2S1471, VIL
Chromosome Location
Chr.2: 218419115 - 218449525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007127.2 NP_009058.2 16 - 17 76
GenBank mRNA AK223398.1 - 16 - 17 76
AK300965.1 - 11 - 12 1266 76
AK313709.1 - 16 - 17 76
X12901.1 - 15 - 16 76

View Full Product Details