Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIL1
Entrez Gene ID
7429
Gene Name
villin 1
Gene Aliases
D2S1471, VIL
Chromosome Location
Chr.2: 218419115 - 218449525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007127.2 NP_009058.2 18 - 19 2292 77
GenBank mRNA AK223398.1 - 18 - 19 2286 77
AK300965.1 - 13 - 14 1523 77
AK313709.1 - 18 - 19 2284 77

View Full Product Details