The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004044.6 | NP_004035.2 | 5 - 6 | 564 | 83 |
XM_017004187.1 | XP_016859676.1 | 5 - 6 | 487 | 83 | |
GenBank mRNA | AB062403.1 | - | 5 - 6 | 412 | 83 |
AK290067.1 | - | 4 - 5 | 546 | 83 | |
AK298142.1 | - | 5 - 6 | 677 | 83 | |
AK313066.1 | - | 5 - 6 | 458 | 83 | |
BC008879.2 | - | 5 - 6 | 407 | 83 | |
D82348.1 | - | 5 - 6 | 452 | 83 | |
D89976.1 | - | 5 - 6 | 434 | 83 | |
DA043224.1 | - | 4 - 5 | 498 | 83 | |
EU794654.1 | - | 5 - 6 | 551 | 83 | |
U37436.1 | - | 4 - 5 | 374 | 83 |