The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004044.6 | NP_004035.2 | 8 - 9 | 998 | 63 |
XM_017004187.1 | XP_016859676.1 | 8 - 9 | 921 | 63 | |
GenBank mRNA | AB062403.1 | - | 8 - 9 | 846 | 63 |
AK290067.1 | - | 7 - 8 | 980 | 63 | |
AK298142.1 | - | 8 - 9 | 1111 | 63 | |
AK313066.1 | - | 8 - 9 | 892 | 63 | |
BC008879.2 | - | 8 - 9 | 841 | 63 | |
D82348.1 | - | 8 - 9 | 886 | 63 | |
D89976.1 | - | 8 - 9 | 868 | 63 | |
EU794654.1 | - | 8 - 9 | 985 | 63 | |
U37436.1 | - | 7 - 8 | 808 | 63 |