The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003728.3 | NP_003719.3 | 2 - 3 | 696 | 87 |
XM_005263321.3 | XP_005263378.1 | 2 - 3 | 696 | 87 | |
XM_017008789.1 | XP_016864278.1 | 3 - 4 | 7113 | 87 | |
XM_017008790.1 | XP_016864279.1 | 2 - 3 | 341 | 87 | |
GenBank mRNA | AF055634.1 | - | 2 - 3 | 497 | 87 |
AK290500.1 | - | 2 - 3 | 507 | 87 | |
AK308421.1 | - | 2 - 3 | 459 | 87 | |
BC041156.1 | - | 2 - 3 | 692 | 87 |