The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044305.2 | NP_001037770.1 | 6 - 7 | 830 | 122 |
NM_001281439.1 | NP_001268368.1 | 5 - 6 | 749 | 122 | |
NM_001281440.1 | NP_001268369.1 | 6 - 7 | 646 | 122 | |
NM_021940.4 | NP_068759.2 | 5 - 6 | 749 | 122 | |
XM_011536045.2 | XP_011534347.1 | 6 - 7 | 807 | 122 | |
XM_011536046.2 | XP_011534348.1 | 7 - 8 | 732 | 122 | |
XM_011536047.2 | XP_011534349.1 | 5 - 6 | 726 | 122 | |
XM_011536048.2 | XP_011534350.1 | 7 - 8 | 719 | 122 | |
XM_017011170.1 | XP_016866659.1 | 7 - 8 | 915 | 122 | |
GenBank mRNA | AF442495.1 | - | 5 - 6 | 647 | 122 |
AK023221.1 | - | 5 - 6 | 564 | 122 | |
AK124239.1 | - | 7 - 8 | 763 | 122 | |
AK222711.1 | - | 5 - 6 | 546 | 122 | |
AK290448.1 | - | 6 - 7 | 607 | 122 | |
AK307807.1 | - | 6 - 7 | 643 | 122 | |
AK315579.1 | - | 5 - 6 | 627 | 122 | |
AY055003.1 | - | 6 - 7 | 645 | 122 | |
AY055004.1 | - | 5 - 6 | 564 | 122 | |
BC028074.1 | - | 5 - 6 | 621 | 122 | |
BC036123.1 | - | 6 - 7 | 683 | 122 |