The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080878.2 | NP_543154.1 | 2 - 3 | 135 | 93 |
XM_011509210.1 | XP_011507512.1 | 4 - 5 | 612 | 93 | |
GenBank mRNA | AY065973.1 | - | 2 - 3 | 107 | 93 |
AY358905.1 | - | 2 - 3 | 135 | 93 |