The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015891.2 | NP_056975.1 | 12 - 13 | 1358 | 135 |
GenBank mRNA | AB208903.1 | - | 12 - 13 | 1332 | 135 |
AF038392.1 | - | 12 - 13 | 1358 | 135 | |
AF061241.1 | - | 12 - 13 | 1331 | 135 | |
AK222721.1 | - | 12 - 13 | 1355 | 135 | |
AK314601.1 | - | 12 - 13 | 1354 | 135 | |
BC111061.1 | - | 12 - 13 | 2764 | 135 | |
BC117145.1 | - | 12 - 13 | 1355 | 135 | |
BC126114.1 | - | 12 - 13 | 1355 | 135 |