The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014390.2 | NP_055205.2 | 2 - 3 | 457 | 64 |
XM_017011987.1 | XP_016867476.1 | 2 - 3 | 486 | 64 | |
GenBank mRNA | AB209510.1 | - | 2 - 3 | 395 | 64 |
AK096583.1 | - | 3 - 4 | 382 | 64 | |
AK304181.1 | - | 2 - 3 | 412 | 64 | |
AK312313.1 | - | 2 - 3 | 411 | 64 | |
BC017180.2 | - | 2 - 3 | 413 | 64 | |
BM149154.1 | - | 2 - 3 | 457 | 64 | |
BT009785.1 | - | 2 - 3 | 231 | 64 | |
U22055.1 | - | 2 - 3 | 423 | 64 |