The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015973.4 | NP_057057.2 | 4 - 5 | 478 | 70 |
GenBank mRNA | BC030241.1 | - | 4 - 5 | 396 | 70 |
M77140.1 | - | 3 - 4 | 172 | 70 |