The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003922.3 | NP_003913.3 | 37 - 38 | 7224 | 66 |
XM_017022699.1 | XP_016878188.1 | 38 - 39 | 7330 | 66 | |
XM_017022700.1 | XP_016878189.1 | 38 - 39 | 7330 | 66 | |
XM_017022701.1 | XP_016878190.1 | 38 - 39 | 7330 | 66 | |
XM_017022702.1 | XP_016878191.1 | 37 - 38 | 7264 | 66 | |
XM_017022703.1 | XP_016878192.1 | 38 - 39 | 7330 | 66 | |
XM_017022704.1 | XP_016878193.1 | 38 - 39 | 7330 | 66 | |
XM_017022705.1 | XP_016878194.1 | 37 - 38 | 7237 | 66 | |
XM_017022706.1 | XP_016878195.1 | 38 - 39 | 7333 | 66 | |
GenBank mRNA | AK226162.1 | - | 24 - 25 | 4287 | 66 |
U50078.1 | - | 37 - 38 | 7172 | 66 |