The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003922.3 | NP_003913.3 | 44 - 45 | 9048 | 70 |
XM_017022699.1 | XP_016878188.1 | 45 - 46 | 9154 | 70 | |
XM_017022700.1 | XP_016878189.1 | 45 - 46 | 9154 | 70 | |
XM_017022701.1 | XP_016878190.1 | 45 - 46 | 9154 | 70 | |
XM_017022702.1 | XP_016878191.1 | 44 - 45 | 9088 | 70 | |
XM_017022703.1 | XP_016878192.1 | 45 - 46 | 9154 | 70 | |
XM_017022705.1 | XP_016878194.1 | 44 - 45 | 9061 | 70 | |
GenBank mRNA | AK226162.1 | - | 31 - 32 | 6111 | 70 |
U50078.1 | - | 44 - 45 | 8996 | 70 |