The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003922.3 | NP_003913.3 | 52 - 53 | 10566 | 63 |
XM_017022699.1 | XP_016878188.1 | 53 - 54 | 10723 | 63 | |
XM_017022700.1 | XP_016878189.1 | 53 - 54 | 10687 | 63 | |
XM_017022701.1 | XP_016878190.1 | 53 - 54 | 10672 | 63 | |
XM_017022702.1 | XP_016878191.1 | 52 - 53 | 10672 | 63 | |
XM_017022703.1 | XP_016878192.1 | 53 - 54 | 10669 | 63 | |
XM_017022704.1 | XP_016878193.1 | 53 - 54 | 10651 | 63 | |
XM_017022705.1 | XP_016878194.1 | 52 - 53 | 10645 | 63 | |
GenBank mRNA | AK226162.1 | - | 39 - 40 | 7629 | 63 |
U50078.1 | - | 52 - 53 | 10514 | 63 |