The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195193.1 | NP_001182122.1 | 5 - 6 | 1474 | 80 |
NM_002482.3 | NP_002473.2 | 7 - 8 | 1666 | 80 | |
NM_152298.3 | NP_689511.2 | 6 - 7 | 649 | 80 | |
XM_005270888.2 | XP_005270945.1 | 8 - 9 | 1686 | 80 | |
XM_005270889.2 | XP_005270946.1 | 7 - 8 | 669 | 80 | |
XM_011541509.1 | XP_011539811.1 | 8 - 9 | 1686 | 80 | |
XM_017001356.1 | XP_016856845.1 | 6 - 7 | 596 | 80 | |
GenBank mRNA | AF035191.1 | - | 6 - 7 | 574 | 80 |
AK056161.1 | - | 8 - 9 | 1682 | 80 | |
AK092829.1 | - | 1 - 2 | 2687 | 80 | |
AK131249.1 | - | 2 - 3 | 1781 | 80 | |
AK222778.1 | - | 7 - 8 | 1609 | 80 | |
AK222816.1 | - | 7 - 8 | 1609 | 80 | |
AK291637.1 | - | 7 - 8 | 1609 | 80 | |
AK298893.1 | - | 5 - 6 | 1417 | 80 | |
AK299584.1 | - | 6 - 7 | 1473 | 80 | |
BC003113.1 | - | 6 - 7 | 567 | 80 | |
BC010105.2 | - | 7 - 8 | 1594 | 80 | |
BC011580.1 | - | 7 - 8 | 1592 | 80 | |
BC053608.1 | - | 6 - 7 | 572 | 80 | |
BT006757.1 | - | 6 - 7 | 490 | 80 | |
M97856.1 | - | 7 - 8 | 1592 | 80 |