The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040107.1 | NP_001035196.1 | 5 - 6 | 601 | 115 |
NM_001256413.1 | NP_001243342.1 | 4 - 5 | 561 | 115 | |
NM_032369.3 | NP_115745.2 | 5 - 6 | 592 | 115 | |
XM_005253948.2 | XP_005254005.1 | 5 - 6 | 679 | 115 | |
XM_011538838.1 | XP_011537140.1 | 6 - 7 | 1017 | 115 | |
XM_011538839.2 | XP_011537141.1 | 6 - 7 | 751 | 115 | |
XM_011538840.2 | XP_011537142.1 | 6 - 7 | 783 | 115 | |
XM_011538841.2 | XP_011537143.1 | 6 - 7 | 761 | 115 | |
XM_011538842.2 | XP_011537144.1 | 6 - 7 | 741 | 115 | |
XM_011538844.2 | XP_011537146.1 | 6 - 7 | 756 | 115 | |
XM_011538845.1 | XP_011537147.1 | 5 - 6 | 636 | 115 | |
XM_011538846.2 | XP_011537148.1 | 6 - 7 | 710 | 115 | |
XM_011538847.1 | XP_011537149.1 | 5 - 6 | 611 | 115 | |
XM_017020026.1 | XP_016875515.1 | 6 - 7 | 725 | 115 | |
XM_017020027.1 | XP_016875516.1 | 5 - 6 | 610 | 115 | |
GenBank mRNA | AK293543.1 | - | 4 - 5 | 527 | 115 |
BC007277.1 | - | 5 - 6 | 1044 | 115 | |
BC009731.1 | - | 5 - 6 | 543 | 115 | |
BC032672.2 | - | 4 - 5 | 1165 | 115 | |
BP369986.1 | - | 5 - 6 | 474 | 115 | |
GU211002.1 | - | 3 - 4 | 411 | 115 |