The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005689.2 | NP_005680.1 | 13 - 14 | 2178 | 70 |
GenBank mRNA | AF070598.1 | - | 9 - 10 | 870 | 70 |
AF076775.1 | - | 13 - 14 | 1861 | 70 | |
AF308472.1 | - | 13 - 14 | 2167 | 70 | |
AJ289233.2 | - | 13 - 14 | 2138 | 70 | |
AK057026.1 | - | 13 - 14 | 1897 | 70 | |
AK172812.1 | - | 13 - 14 | 1528 | 70 | |
BC000559.2 | - | 13 - 14 | 2129 | 70 | |
BC043423.1 | - | 6 - 7 | 2178 | 70 | |
U66673.1 | - | 2 - 3 | 154 | 70 |