The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293163.1 | NP_001280092.1 | 10 - 11 | 1478 | 63 |
GenBank mRNA | BC016925.1 | - | 10 - 11 | 1424 | 63 |
EU159452.1 | - | 9 - 10 | 1347 | 63 |