The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243900.2 | NP_001230829.1 | 19 - 20 | 2762 | 96 |
NM_001320965.1 | NP_001307894.1 | 19 - 20 | 2903 | 96 | |
NM_001320966.1 | NP_001307895.1 | 19 - 20 | 2761 | 96 | |
NM_001320967.1 | NP_001307896.1 | 19 - 20 | 2838 | 96 | |
NM_005336.5 | NP_005327.1 | 19 - 20 | 2985 | 96 | |
NM_203346.4 | NP_976221.1 | 19 - 20 | 2838 | 96 | |
XM_005247002.3 | XP_005247059.2 | 19 - 20 | 2792 | 96 | |
XM_005247003.4 | XP_005247060.2 | 19 - 20 | 2837 | 96 | |
XM_006712475.3 | XP_006712538.1 | 21 - 22 | 3041 | 96 | |
XM_011511058.2 | XP_011509360.1 | 21 - 22 | 3205 | 96 | |
XM_011511060.2 | XP_011509362.1 | 20 - 21 | 3053 | 96 | |
XM_017003940.1 | XP_016859429.1 | 20 - 21 | 2863 | 96 | |
XM_017003941.1 | XP_016859430.1 | 19 - 20 | 2980 | 96 | |
XM_017003942.1 | XP_016859431.1 | 19 - 20 | 2863 | 96 | |
GenBank mRNA | AK127833.1 | - | 13 - 14 | 1942 | 96 |
AK131375.1 | - | 8 - 9 | 2195 | 96 | |
AK296684.1 | - | 13 - 14 | 1783 | 96 | |
AK300312.1 | - | 19 - 20 | 2707 | 96 | |
AK312335.1 | - | 19 - 20 | 2815 | 96 | |
BC001179.1 | - | 19 - 20 | 2811 | 96 | |
BC014305.2 | - | 11 - 12 | 1500 | 96 | |
BC113120.1 | - | 3 - 4 | 575 | 96 | |
BX647759.1 | - | 19 - 20 | 2846 | 96 | |
M64098.1 | - | 19 - 20 | 2764 | 96 |