The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243900.2 | NP_001230829.1 | 20 - 21 | 2899 | 87 |
NM_001320965.1 | NP_001307894.1 | 20 - 21 | 3040 | 87 | |
NM_001320966.1 | NP_001307895.1 | 20 - 21 | 2898 | 87 | |
NM_001320967.1 | NP_001307896.1 | 20 - 21 | 2975 | 87 | |
NM_005336.5 | NP_005327.1 | 20 - 21 | 3122 | 87 | |
NM_203346.4 | NP_976221.1 | 20 - 21 | 2975 | 87 | |
XM_005247002.3 | XP_005247059.2 | 20 - 21 | 2929 | 87 | |
XM_005247003.4 | XP_005247060.2 | 20 - 21 | 2974 | 87 | |
XM_006712475.3 | XP_006712538.1 | 22 - 23 | 3178 | 87 | |
XM_011511058.2 | XP_011509360.1 | 22 - 23 | 3342 | 87 | |
XM_011511060.2 | XP_011509362.1 | 21 - 22 | 3190 | 87 | |
XM_017003940.1 | XP_016859429.1 | 21 - 22 | 3000 | 87 | |
XM_017003941.1 | XP_016859430.1 | 20 - 21 | 3117 | 87 | |
XM_017003942.1 | XP_016859431.1 | 20 - 21 | 3000 | 87 | |
GenBank mRNA | AK127833.1 | - | 14 - 15 | 2079 | 87 |
AK131375.1 | - | 9 - 10 | 2332 | 87 | |
AK296684.1 | - | 14 - 15 | 1920 | 87 | |
AK312335.1 | - | 20 - 21 | 2952 | 87 | |
BC001179.1 | - | 20 - 21 | 2948 | 87 | |
BC014305.2 | - | 12 - 13 | 1637 | 87 | |
BC113120.1 | - | 4 - 5 | 712 | 87 | |
BX647759.1 | - | 20 - 21 | 2983 | 87 | |
M64098.1 | - | 20 - 21 | 2901 | 87 |