Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDLBP
Entrez Gene ID
3069
Gene Name
high density lipoprotein binding protein
Gene Aliases
HBP, PRO2900, VGL
Chromosome Location
Chr.2: 241227264 - 241315842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320965.1 NP_001307894.1 2 - 3 261 66
NM_001320966.1 NP_001307895.1 2 - 3 119 66
NM_001320967.1 NP_001307896.1 2 - 3 196 66
NM_005336.5 NP_005327.1 2 - 3 343 66
NM_203346.4 NP_976221.1 2 - 3 196 66
XM_005247002.3 XP_005247059.2 2 - 3 150 66
XM_005247003.4 XP_005247060.2 2 - 3 195 66
XM_006712475.3 XP_006712538.1 4 - 5 399 66
XM_011511058.2 XP_011509360.1 4 - 5 563 66
XM_011511060.2 XP_011509362.1 3 - 4 411 66
XM_017003940.1 XP_016859429.1 3 - 4 221 66
XM_017003941.1 XP_016859430.1 2 - 3 338 66
XM_017003942.1 XP_016859431.1 2 - 3 221 66
GenBank mRNA AK296684.1 - 2 - 3 173 66
AK309075.1 - 2 - 3 152 66
AL701577.1 - 2 - 3 146 66
BC001179.1 - 2 - 3 169 66
BG720303.1 - 2 - 3 345 66
BP283713.1 - 2 - 3 119 66
BP359370.1 - 2 - 3 196 66
BX647759.1 - 2 - 3 204 66
DB282620.1 - 2 - 3 189 66
DC400438.1 - 2 - 3 261 66
DC424146.1 - 2 - 3 343 66
M64098.1 - 2 - 3 122 66

View Full Product Details