The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025184.3 | NP_079460.2 | 3 - 4 | 468 | 107 |
XM_006724562.2 | XP_006724625.1 | 2 - 3 | 181 | 107 | |
GenBank mRNA | AK026496.1 | - | 3 - 4 | 321 | 107 |
AK291187.1 | - | 3 - 4 | 609 | 107 | |
AK292967.1 | - | 3 - 4 | 452 | 107 | |
BC031039.1 | - | 3 - 4 | 463 | 107 |