Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
Chromosome Location
Chr.X: 44146254 - 44343677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025184.3 NP_079460.2 9 - 10 1513 86
XM_006724562.2 XP_006724625.1 8 - 9 1226 86
GenBank mRNA AK026496.1 - 9 - 10 1367 86
AK291187.1 - 9 - 10 1654 86
AK292967.1 - 9 - 10 1497 86
BC031039.1 - 9 - 10 1508 86
CR749363.1 - 4 - 5 1174 86

View Full Product Details