The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020372.3 | NP_065105.2 | 7 - 8 | 1454 | 58 |
XM_005267747.4 | XP_005267804.1 | 8 - 9 | 3797 | 58 | |
XM_017021361.1 | XP_016876850.1 | 8 - 9 | 3794 | 58 | |
GenBank mRNA | BC020565.1 | - | 7 - 8 | 1273 | 58 |