The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181701.3 | NP_859052.3 | 6 - 7 | 866 | 77 |
GenBank mRNA | AJ318051.1 | - | 6 - 7 | 866 | 77 |
AK128077.1 | - | 6 - 7 | 531 | 77 | |
AL834369.1 | - | 3 - 4 | 357 | 77 | |
BC047604.2 | - | 7 - 8 | 748 | 77 |