The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278541.1 | NP_001265470.1 | 8 - 9 | 953 | 146 |
NM_001872.4 | NP_001863.3 | 9 - 10 | 1064 | 146 | |
XM_017020393.1 | XP_016875882.1 | 9 - 10 | 1007 | 146 | |
GenBank mRNA | AB011969.1 | - | 8 - 9 | 905 | 146 |
AK290829.1 | - | 9 - 10 | 1012 | 146 | |
BC007057.1 | - | 9 - 10 | 1021 | 146 | |
BT006936.1 | - | 9 - 10 | 997 | 146 | |
M75106.1 | - | 9 - 10 | 1016 | 146 |