The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000189.4 | NP_000180.2 | 17 - 18 | 4531 | 65 |
XM_005264280.1 | XP_005264337.1 | 17 - 18 | 3074 | 65 | |
XM_011532807.1 | XP_011531109.1 | 16 - 17 | 2978 | 65 | |
XM_017003945.1 | XP_016859434.1 | 16 - 17 | 2915 | 65 | |
GenBank mRNA | AF148513.1 | - | 17 - 18 | 3054 | 65 |
AK290357.1 | - | 17 - 18 | 3034 | 65 | |
BC021116.1 | - | 17 - 18 | 3049 | 65 | |
BC064369.1 | - | 17 - 18 | 3047 | 65 | |
CR749202.1 | - | 17 - 18 | 2711 | 65 |